Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Sessole chimiche

Sessole chimiche

2580 ml, 430 mm
Catalog # RSGA748.430
Supplier:   RSG
Sessole chimiche
Catalog # RSGA748.430
Supplier:  RSG
Sessole chimiche
Sessole chimiche

2580 ml, 430 mm

Price
SFr. 119.00
Log in for availability
1 * 1 ST
Product Details & Documents

These chemical scoops are made from aluminium.

Documents

Specifications
  • Capacità:
    2580 ml
  • Lunghezza:
    430 mm
  • Seq:
    09
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.