Membrane filtration set
Membrane filtration set including 50 syringe filters and 1 disposable 10 ml syringe, 1,20 µm
Codice catalogo HACHLCW904
Nome del fornitore: Hach
Vai a
Product Details & Documents
This membrane filtration set can be used for filtration of turbid samples for the determination of dissolved ingredients.
Delivery information: The set contains 50 syringe filters and one disposable 10 ml syringe.
Specifiche
- Dimensioni pori:1,20 µm
- Descrizione:Membrane filtration set including 50 syringe filters and 1 disposable 10 ml syringe
- Seq:02