Sample tubes
0,5 ml, Conical bottom, trasparente, without side label, PP, screw cap with attachment loop, Sterili
Codice catalogo BZYM710061
Marketsource
Nome del fornitore: BIOZYM
Vai a
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Specifiche
- Sterilità:Sterili
- Tipo:screw cap with attachment loop
- Capacità:0,5 ml
- Colore:trasparente
- Descrizione:without side label
- Materiale:PP
- Fondo:Conical bottom