Sample tubes
2,0 ml, Conical bottom, trasparente, with side label, PP, screw cap without attachment loop, Sterili
Codice catalogo BZYM710053
Marketsource
Nome del fornitore: BIOZYM
Vai a
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Specifiche
- Sterilità:Sterili
- Tipo:screw cap without attachment loop
- Capacità:2,0 ml
- Colore:trasparente
- Descrizione:with side label
- Materiale:PP
- Seq:21
- Fondo:Conical bottom