Sample tubes
0,5 ml, Conical bottom, trasparente, without side label, PP, screw cap without attachment loop, Sterili
Catalog # BZYM710051
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Specifications
- Sterilità:Sterili
- Tipo:screw cap without attachment loop
- Capacità:0,5 ml
- Colore:trasparente
- Descrizione:without side label
- Materiale:PP
- Seq:13
- Fondo:Conical bottom