Sample tubes
0,5 ml, Free standing, ambra, with side label, PP
Codice catalogo BZYM710016
Marketsource
Nome del fornitore: BIOZYM
Vai a
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
With or without screw caps.
Specifiche
- Capacità:0,5 ml
- Colore:ambra
- Descrizione:with side label
- Materiale:PP
- Fondo:Free standing