Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Cryogenic vials, Cryovials®

10,00 ml, Freestanding, 17,0×84 mm, Yes
  BZYM710518
Marketsource
 :   BIOZYM
Cryogenic vials, Cryovials®
  BZYM710518
 :  BIOZYM
Marketsource
Cryogenic vials, Cryovials®

10,00 ml, Freestanding, 17,0×84 mm, Yes

 
SFr. 23.82
   
1 * 50 ST
 

These cryogenic vials made from PP are used for optimum storage of a variety of biological materials such as cell cultures, sera and blood components.

  • Gamma sterilised and DNA-, DNAse-, RNAse- and pyrogen free
  • Silicone sealing ring for safe and dense closure
  • Vertical lids for tight fingers
  • White, scratch-resistant label
  • Perfect sample visibility through clear vascular walls
  • Snap-in thread for all free-standing tubes

Documents

 
  • Tappo incluso:
    Yes
  • Ø×A:
    17,0×84 mm
  • Capacità:
    10,00 ml
  • Seq:
    13
  • Fondo:
    Freestanding
Accessories

 

  •  
    Descrizione
     
     
  • BZYM710550
    Aluminium stand for up to 5 tubes of sizes 1 and 2 ml
       
    1 * 12 ST - SFr. 16.03

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.