Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Cryogenic vials, Cryovials®

Cryogenic vials, Cryovials®

2,00 ml, Conical bottom, 12,5×49 mm, Yes
  BZYM710502
Marketsource
 :   BIOZYM
Cryogenic vials, Cryovials®
  BZYM710502
 :  BIOZYM
Marketsource
Cryogenic vials, Cryovials®

2,00 ml, Conical bottom, 12,5×49 mm, Yes

 
SFr. 29.32
   
1 * 100 ST
 

These cryogenic vials made from PP are used for optimum storage of a variety of biological materials such as cell cultures, sera and blood components.

  • Gamma sterilised and DNA-, DNAse-, RNAse- and pyrogen free
  • Silicone sealing ring for safe and dense closure
  • Vertical lids for tight fingers
  • White, scratch-resistant label
  • Perfect sample visibility through clear vascular walls
  • Snap-in thread for all free-standing tubes

Documents

 
  • Tappo incluso:
    Yes
  • Ø×A:
    12,5×49 mm
  • Capacità:
    2,00 ml
  • Seq:
    02
  • Fondo:
    Conical bottom
Accessories

 

  •  
    Descrizione
     
     
  • BZYM710550
    Aluminium stand for up to 5 tubes of sizes 1 and 2 ml
       
    1 * 12 ST - SFr. 16.03