VISOCOLOR® Colorimetric Test, Hardness
VISOCOLOR® HE Total hardness H 20 F, Reagents for 200 tests, titration syringe, test tube with ring mark and 2 plastic dropping tips, 0,6 - 25,0 °e (1 syringe filling); 0,1 - 3,6 mmol/L (Ca²⁺) (1 syringe filling), Phenolphthalein, Visual, 170×125×45 mm (Including packaging), 200, 1 KIT
Reagents for the determination of hardness in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
The term carbonate hardness describes the amount of magnesium and calcium ions for which equivalent molar amounts of hydrogen carbonate are available in the solution.
Hazardous material: Yes
- Metodo chimico:Phenolphthalein
- Conf:1 KIT
- Descrizione:VISOCOLOR® HE Total hardness H 20 F
- Dimensioni:170×125×45 mm (Including packaging)
- Per strumento:Visual
- Test per kit:200
- Range di misura:0,6 - 25,0 °e (1 syringe filling); 0,1 - 3,6 mmol/L (Ca²⁺) (1 syringe filling)
- Indice:Reagents for 200 tests, titration syringe, test tube with ring mark and 2 plastic dropping tips
- Cat. No.:MANA915005
- Peso:265,5 g (Including packaging)
- Piattaforma:Titrimetric test kits
- Durata a magazzino (mesi):18
- Temperatura di conservazione:15...25 °C
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



