VISOCOLOR® Colorimetric Test, Hardness
VISOCOLOR® alpha Total hardness, Reagents for 100 tests in a blister, 1 drop corresponds to 1 - °e, Complexometric titration, Visual, 150×93×35 mm (Including packaging), 100, 1 KIT
Nome del fornitore: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Hardness
Codice catalogo MANA935042
Nome del fornitore: MACHEREY-NAGEL
Numero CAS:
Vai a
Product Details & Documents
Reagents for the determination of hardness in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
The term carbonate hardness describes the amount of magnesium and calcium ions for which equivalent molar amounts of hydrogen carbonate are available in the solution.
Hazardous material: Yes
Specifiche
Specifiche
- Metodo chimico:Complexometric titration
- Conf:1 KIT
- Descrizione:VISOCOLOR® alpha Total hardness
- Dimensioni:150×93×35 mm (Including packaging)
- Per strumento:Visual
- Test per kit:100
- Range di misura:1 drop corresponds to 1 - °e
- Indice:Reagents for 100 tests in a blister
- Cat. No.:MANA935042
- Peso:64 g (Including packaging)
- Piattaforma:Titrimetric test kits
- Durata a magazzino (mesi):18
- Temperatura di conservazione:15...25 °C
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



