Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

VISOCOLOR® Colorimetric Test, Chromium

VISOCOLOR® ECO Chromium(VI), refill pack, test no. 5-20, Reagents for 140 tests in a box, 0,02 • 0,05 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,50 mg/L; 0,02 - 0,50 mg/L, Carbazide, PF-12Plus; Advance; Visual, 170×115×70 mm (Including packaging), 140, 1 KIT
Catalog # MANA931220
Supplier:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Chromium
Catalog # MANA931220
Supplier:  MACHEREY-NAGEL
CAS Number:  
VISOCOLOR® Colorimetric Test, Chromium
VISOCOLOR® Colorimetric Test, Chromium

VISOCOLOR® ECO Chromium(VI), refill pack, test no. 5-20, Reagents for 140 tests in a box, 0,02 • 0,05 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,50 mg/L; 0,02 - 0,50 mg/L, Carbazide, PF-12Plus; Advance; Visual, 170×115×70 mm (Including packaging), 140, 1 KIT

CAS Number:  
Price
SFr. 54.50
Log in for availability
1 * 1 KIT
Product Details & Documents

Reagents for the determination of chromium in water samples.

  • Combines simple implementation with high measurement reliability

Simple determination directly on site or in the laboratory.

Sea water analysis: Yes

Remark: Measuring range on PF-12Plus. Range on other photometers can be different.

Hazardous material: Yes

Specifications

Specifications

  • Metodo chimico:
    Carbazide
  • Conf:
    1 KIT
  • Descrizione:
    VISOCOLOR® ECO Chromium(VI), refill pack, test no. 5-20
  • Dimensioni:
    170×115×70 mm (Including packaging)
  • Per strumento:
    PF-12Plus; Advance; Visual
  • Test per kit:
    140
  • Range di misura:
    0,02 • 0,05 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,50 mg/L; 0,02 - 0,50 mg/L
  • Indice:
    Reagents for 140 tests in a box
  • Cat. No.:
    MANA931220
  • Peso:
    149,5 g (Including packaging)
  • Modello:
    MANA931220
  • Piattaforma:
    Colorimetric test kits
  • Durata a magazzino (mesi):
    18
  • Temperatura di conservazione:
    15...25 °C

Specifications