VISOCOLOR® Colorimetric Test, Hardness
VISOCOLOR® ECO Total hardness, Reagents for 110 tests and accessories in a box, 1 drop corresponds to 1 °e, Complexometric titration, Visual, 170×125×48 mm (Including packaging), 110, 1 KIT
Reagents for the determination of hardness in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
The term carbonate hardness describes the amount of magnesium and calcium ions for which equivalent molar amounts of hydrogen carbonate are available in the solution.
Hazardous material: Yes
- Metodo chimico:Complexometric titration
- Conf:1 KIT
- Descrizione:VISOCOLOR® ECO Total hardness
- Dimensioni:170×125×48 mm (Including packaging)
- Per strumento:Visual
- Test per kit:110
- Range di misura:1 drop corresponds to 1 °e
- Indice:Reagents for 110 tests and accessories in a box
- Cat. No.:MANA931029
- Peso:202,2 g (Including packaging)
- Piattaforma:Titrimetric test kits
- Durata a magazzino (mesi):18
- Temperatura di conservazione:15...25 °C