VISOCOLOR® Colorimetric Test, Chloride
VISOCOLOR® ECO Chloride, test no. 5-18. Measuring range on PF-12Plus. Range on other photometers can be different, Reagents for 90 tests, color chart and accessories in a box, 1 • 2 • 4 • 7 • 12 • 20 • 40 • 60 mg/L (Cl⁻); 1 - 50 mg/L (Cl⁻), Mercury(II), PF-12Plus;Advance;Visual, 170×125×48 mm (Including packaging), Sea water non compatible, 90, 1 KIT
Nome del fornitore: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Chloride
Codice catalogo MANA931018
Nome del fornitore: MACHEREY-NAGEL
Numero CAS:
Vai a
Product Details & Documents
Reagents for the determination of chloride in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Specifiche
Specifiche
- Metodo chimico:Mercury(II)
- Conf:1 KIT
- Descrizione:VISOCOLOR® ECO Chloride, test no. 5-18. Measuring range on PF-12Plus. Range on other photometers can be different
- Dimensioni:170×125×48 mm (Including packaging)
- Tipo:Sea water non compatible
- Per strumento:PF-12Plus;Advance;Visual
- Test per kit:90
- Range di misura:1 • 2 • 4 • 7 • 12 • 20 • 40 • 60 mg/L (Cl⁻); 1 - 50 mg/L (Cl⁻)
- Indice:Reagents for 90 tests, color chart and accessories in a box
- Cat. No.:MANA931018
- Peso:252 g (Including packaging)
- Modello:MANA931018
- Piattaforma:Colorimetric test kits
- Durata a magazzino (mesi):12
- Temperatura di conservazione:15...25 °C
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



