SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
NANO Reagent for lime precipitation 1 * 1 KIT
MANA918939
:
MACHEREY-NAGEL
NANO Reagent for lime precipitation 1 * 1 KIT
MANA918939
:
MACHEREY-NAGEL
IFU_Nano Reagent For Lime Precipitation.pdf
SDS_Nano Reagent For Lime Precipitation.pdf
Frequently Bought Together
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Salviette per il viso
: VWR Collection
VWR®, Guanti in Nitrile
: VWR Collection
Strisce reattive pH, MQuant®, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Guanti in nitrile
: VWR Collection
Acetonitrile, anidro (max. 0,003% H<sub>2</sub>O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade adatto per UPLC/UHPLC
: VWR Chemicals
VWR®, pipette di trasferimento, alte prestazioni
: VWR Collection
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., grado gradiente for liquid chromatography, Supelco®
: Merck
Acqua, HiPerSolv CHROMANORM® per HPLC
: VWR Chemicals
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
: VWR Chemicals
Guanti in nitrile, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Pipette sierologiche, Standard Line
: VWR Collection