VISOCOLOR® Colorimetric Test, Hardness
VISOCOLOR® HE Total hardness H 20 F, refill pack, Reagents for 200 tests in a box, 0,6 - 25,0 °e (1 syringe filling); 0,1 - 3,6 mmol/L (Ca²⁺) (1 syringe filling), Complexometric titration, Visual, 170×115×70 mm (Including packaging), 200, 1 KIT
Nome del fornitore: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Hardness
Codice catalogo MANA915205
Nome del fornitore: MACHEREY-NAGEL
Numero CAS:
Vai a
Product Details & Documents
Reagents for the determination of hardness in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
The term carbonate hardness describes the amount of magnesium and calcium ions for which equivalent molar amounts of hydrogen carbonate are available in the solution.
Hazardous material: Yes
Specifiche
Specifiche
- Metodo chimico:Complexometric titration
- Conf:1 KIT
- Descrizione:VISOCOLOR® HE Total hardness H 20 F, refill pack
- Dimensioni:170×115×70 mm (Including packaging)
- Per strumento:Visual
- Test per kit:200
- Range di misura:0,6 - 25,0 °e (1 syringe filling); 0,1 - 3,6 mmol/L (Ca²⁺) (1 syringe filling)
- Indice:Reagents for 200 tests in a box
- Cat. No.:MANA915205
- Peso:197,8 g (Including packaging)
- Piattaforma:Titrimetric test kits
- Durata a magazzino (mesi):18
- Temperatura di conservazione:15...25 °C



