Empty solid phase extraction columns, CHROMABOND®
Empty PP columns with PE frits, one filter element is already inserted in the PP column, 6 ml
Catalog # MANA730161
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Empty and reservoir columns with accessories for solid phase extraction.
- Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
- Reservoir columns from PP for application of medium-size samples
- Accessories for SPE columns (filter elements, adapters)
Specifications
- Capacità:6 ml
- Descrizione:Empty PP columns with PE frits, one filter element is already inserted in the PP column
- Seq:03
Accessories
Product Information
- Catalog #DescrizioneAvailabilityPrice
- MANA732019PE frits for LV PP columns 15 ml, for 100 mg adsorbent weightLog in for availability1 * 250 ST - SFr. 147.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- MANA732020PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weightLog in for availability1 * 250 ST - SFr. 206.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- MANA730101Adapters (PP) for PP columns (1, 3 and 6 ml)Log in for availability1 * 10 ST - SFr. 107.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- MANA730105Adapters (PVDF) for glass columns (3 and 6 ml)Log in for availability1 * 10 ST - SFr. 223.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
- MANA730192Glass fiber frits for glass columns, 6 mlLog in for availability1 * 250 ST - SFr. 265.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
- MANA730566Adapters (PE) for PP columns (30 and 70 ml)Log in for availability1 * 1 KIT - SFr. 32.40
Product Information
- Catalog #DescrizioneAvailabilityPrice
- MANA730385Adapters (PE) for PP columns (15, 45 and 70 ml)Log in for availability1 * 10 ST - SFr. 139.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- MANA730191Glass fiber frits for glass columns, 3 mlLog in for availability1 * 250 ST - SFr. 218.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice