Snap ring caps for snap ring vials, N 11
PE, hard, transparent, center hole, 50° shore A, PTFE red/silicone white/PTFE red, 1,0 mm
Codice catalogo MANA702718
Nome del fornitore: MACHEREY-NAGEL
Snap ring caps for snap ring vials, N 11
Codice catalogo MANA702718
Nome del fornitore: MACHEREY-NAGEL
Vai a
Product Details & Documents
Ready assembled snap ring closures made of polyethylene are available for the snap ring vials.
- Available as hard cap version in transparent and further colours or as soft cap in light blue for easy handling
- Large choice of septa materials
- Most frequent version are closures with cross-slit silicone/PTFE septum, which supports easy penetration of blunt HPLC needles
Documents
MACHEREY-NAGEL Vials and Caps.pdf
Ulteriori dettagli sul prodotto
Specifiche
- Durezza:50° shore A
- Setto:PTFE red/silicone white/PTFE red
- Tipo:PE, hard, transparent, center hole
- Spessore:1,0 mm
- Cat. No.:MANA702718



