Orders LinkContactUsLinkComponent
Glicina
Glicina
Bottiglia di vetro, 1 kg
Catalog # A1067.1000
Supplier:   PanReac AppliChem
Glicina
Catalog # A1067.1000
Supplier:  PanReac AppliChem
CAS Number:  56-40-6
Glicina
Glicina
Bottiglia di vetro, 1 kg
CAS Number:  56-40-6
Price
SFr. 112.00
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Aminoacetic acid, Glycocoll, H-Gly-OH
  • CAS Number:56-40-6
  • Formula:NH₂CH₂COOH
  • MDL Number:MFCD00008131
  • Molecular Weight:75.07 g/mol
  • Merck Index:13,04504
  • Boiling Pt:181 °C (1013 hPa)
  • Melting Pt:83 °C
  • Density:1,64 g/cm³ (20 °C)
  • Storage Temperature:Ambiente
  • EINECS:200-272-2
Documents
Specifications
Specifications
  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    1 kg
Specifications