Fmoc-Pro-OH
Glass bottle for solids, 100 g
Codice catalogo BACE4003150.0100
Marketsource
Nome del fornitore: BACHEM BIOCHEMICA
Fmoc-Pro-OH
Codice catalogo BACE4003150.0100
Nome del fornitore: BACHEM BIOCHEMICA
Marketsource
Numero CAS: 71989-31-6
Vai a
Product Details & Documents
Sinonimi: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline
- Numero CAS:71989-31-6
- Formula:C₂₀H₁₉NO₄
- Numero MDL:MFCD00037122
- Molecular Weight:337.38 g/mol
- EINECS:276-259-0
Specifiche
Specifiche
- Tipo di conf.:Glass bottle for solids
- Conf:100 g
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



