Orders LinkContactUsLinkComponent
D-(+)-Tyrosine
Glass bottle for solids, 25 g
  BACE4026808.0025
Marketsource
D-(+)-Tyrosine
  BACE4026808.0025
Marketsource
 :  556-02-5
D-(+)-Tyrosine
Glass bottle for solids, 25 g
 :  556-02-5
1 * 25 g
 :  H-D-Tyr-OH
  •  :556-02-5
  •  :C₉H₁₁NO₃
  •  :MFCD00063073
  •  :181.19 g/mol
  •  :12,09970
  •  :351 °C (1013 hPa)
  •  :285 °C
  •  :209-112-6
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    25 g