VISOCOLOR® Colorimetric Test, Chromium
VISOCOLOR® ECO Chromium(VI), test no. 5-20, Reagents for 140 tests, colour chart and accessories in a box, 0,02 • 0,05 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,50 mg/L; 0,02 - 0,50 mg/L, Carbazide, PF-12Plus; PF-3 Fish; Advance; Visual, 168×112×40 mm (Including packaging), 140, 1 KIT
Nome del fornitore: MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Chromium
Codice catalogo MANA931020
Nome del fornitore: MACHEREY-NAGEL
Numero CAS:
Vai a
Product Details & Documents
Reagents for the determination of chromium in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Remark: Measuring range on PF-12Plus. Range on other photometers can be different.
Hazardous material: Yes
Specifiche
Specifiche
- Metodo chimico:Carbazide
- Conf:1 KIT
- Descrizione:VISOCOLOR® ECO Chromium(VI), test no. 5-20
- Dimensioni:168×112×40 mm (Including packaging)
- Per strumento:PF-12Plus; PF-3 Fish; Advance; Visual
- Test per kit:140
- Range di misura:0,02 • 0,05 • 0,10 • 0,15 • 0,20 • 0,30 • 0,40 • 0,50 mg/L; 0,02 - 0,50 mg/L
- Indice:Reagents for 140 tests, colour chart and accessories in a box
- Cat. No.:MANA931020
- Peso:239,4 g (Including packaging)
- Modello:MANA931020
- Piattaforma:Colorimetric test kits
- Durata a magazzino (mesi):18
- Temperatura di conservazione:15...25 °C
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



