Erlenmeyer Flasks with screw cap
2000 ml, GL 45
Codice catalogo NEUB0748-10-2000
Nome del fornitore: NEUBERT VOLUME GLASSWAERE
Erlenmeyer Flasks with screw cap
Codice catalogo NEUB0748-10-2000
Nome del fornitore: NEUBERT VOLUME GLASSWAERE
Vai a
Product Details & Documents
Borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area. Screw cap with PTFE coated seal.
Specifiche
- Filettatura:GL 45
- Capacità:2000 ml
- Seq:02