L(-)-Glutatione (forma ridotta) ≥98%, purissimo
Bottiglia di plastica, 250 g
Supplier: VWR Chemicals
L(-)-Glutatione (forma ridotta) ≥98%, purissimo
Catalog # 0399-250G
Supplier: VWR Chemicals
CAS Number: 70-18-8
Jump to:
Product Details & Documents
Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
- CAS Number:70-18-8
- Formula:C₁₀H₁₇N₃O₆S
- MDL Number:MFCD00065939
- Molecular Weight:307.33 g/mol
- Merck Index:13,04488
- Melting Pt:192…195 °C
- Melting Pt:192−195 °C
- Density:1,475 g/cm³ (20 °C)
- Storage Temperature:Ambiente
- EINECS:200-725-4
Documents
Specifications
Specification Test Results
- Iron:≤0.002%
- Lead:≤0.001%
- Loss on drying:≤1%
- Purity:≥98%
- Specific rotation:-18.5 ° a -15.5 °
Specifications
- Tipo di conf.:Bottiglia di plastica
- Conf:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



