Flaconi spray
400 ml
Codice catalogo 215-2320
Nome del fornitore: Brand
Vai a
Product Details & Documents
HDPE, white.
- Good chemical resistance
- Good temperature resistance
- Robust, convenient, stable
By turning the nozzle, delivery can be varied from a fine spray to a solid stream.
Specifiche
- Capacità:400 ml
- Seq:01