Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Cryogenic vial boxes, Cryule®, WHEATON®

Cryogenic vial boxes, Cryule®, WHEATON®

81, Fiale Cryule® 2 ml, 133×133×44 mm
Codice catalogo WHEA651492
Nome del fornitore:   DWK Life Sciences
Cryogenic vial boxes, Cryule®, WHEATON®
Codice catalogo WHEA651492
Nome del fornitore:  DWK Life Sciences
Cryogenic vial boxes, Cryule®, WHEATON®
Cryogenic vial boxes, Cryule®, WHEATON®

81, Fiale Cryule® 2 ml, 133×133×44 mm

Price
SFr. 230.00
Log in per disponibilità
1 * 15 ST
Product Details & Documents

These cryoboxes are made of high quality chip-board and spray coated with polyethylene.

  • Stores 1 and 2 ml cryogenic vials
  • Holds 81 freestanding or round bottom vials

Suitable for use with vapor phase of liquid nitrogen.

Specifiche
  • Scomparti:
    81
  • Per:
    Fiale Cryule® 2 ml
  • Esterno L×P×A:
    133×133×44 mm