Scintillation vials, PET, WHEATON®
20,00 ml, 27×61* mm, Yes, Urea, Poly-seal cone
Codice catalogo 548-2049
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Manufactured from clear polyethylene terephthalate (PET) these vials have advantages over glass counting vials of low background count, economy and virtual unbreakability.
- Clarity of glass
- Can be safely incinerated
- Lightweight and virtually unbreakable
* With cap on
Specifiche
- Tappo incluso:Yes
- Materiale tappo:Urea
- Ø×A:27×61* mm
- Setto:Poly-seal cone
- Capacità:20,00 ml