Drum thieves, Wheaton®
Drum thieves, Vetro, 25 ml
Codice catalogo WHEA885300
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Drum thieves made of soda lime glass for quick disposal.
- Conventional open end drum thieves
- Just insert empty tube in drum
- Hold thumb over top and withdraw
Specifiche
- Capacità:25 ml
- Descrizione:Drum thieves
- Materiale:Vetro