Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
EPA vials with screw cap, WHEATON®

EPA vials with screw cap, WHEATON®

40,00 ml, 28×98 mm, ambra, Yes, Screw cap, open top, 24-400, PP, bianco, PTFE faced silicone
Codice catalogo WHEA225315
Nome del fornitore:   DWK Life Sciences
EPA vials with screw cap, WHEATON®
Codice catalogo WHEA225315
Nome del fornitore:  DWK Life Sciences
EPA vials with screw cap, WHEATON®
EPA vials with screw cap, WHEATON®

40,00 ml, 28×98 mm, ambra, Yes, Screw cap, open top, 24-400, PP, bianco, PTFE faced silicone

Price
SFr. 491.00
Log in per disponibilità
1 * 72 ST
Product Details & Documents

Clear vials are manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E438 Type I, Class A requirements. Amber vials are manufactured from WHEATON 320 amber glass that conforms to USP Type I requirements.

  • Amber vials protect light-sensitive analytes from break-down
  • Clear vial with open top black phenolic PTFE faced silicone lined cap
  • Amber vial with open top white polypropylene PTFE faced silicone lined cap
  • Closures are assembled to vials

The EPA Vial is ideal for use in water sampling according to EPA 40 CFR 136, 'Guidelines for Establishing Test Procedures for the Analysis of Pollutants'.

Specifiche
  • Tappo incluso:
    Yes
  • Materiale tappo:
    PP
  • Dimensione tappo:
    24-400
  • Ø×A:
    28×98 mm
  • Setto:
    PTFE faced silicone
  • Capacità:
    40,00 ml
  • Colore:
    ambra
  • Seq:
    02
  • Tipologia tappo:
    Screw cap, open top
  • Colore tappo:
    bianco