BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 361-384, 300 ml
WHEA227497-16G
Made of Type I borosilicate glass.
- Available with or without stopper
- With white labeling area
- Capacità:300 ml
- Descrizione:BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 361-384