Orders LinkContactUsLinkComponent
BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 337-360, 300 ml
Codice catalogo WHEA227497-15
Nome del fornitore:   DWK Life Sciences
BOD bottles, WHEATON®
Codice catalogo WHEA227497-15
Nome del fornitore:  DWK Life Sciences
BOD bottles, WHEATON®
BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 337-360, 300 ml
Price
SFr. 854.00
Log in per disponibilità
1 * 24 ST
Product Details & Documents
Made of Type I borosilicate glass.
  • Available with or without stopper
  • With white labeling area
Specifiche
  • Capacità:
    300 ml
  • Descrizione:
    BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 337-360