Spray bottles, WHEATON®
60 ml, 108 mm, con tappo a vite in PP
Codice catalogo WHEA226436
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Bottle made of white LDPE (Low Density Polyethylene).
- Includes bottle, atomizer tip, dip tube and screw cap
- Screw cap made from polypropylene
Oval spray bottle.
Specifiche
- Tipo:con tappo a vite in PP
- Capacità:60 ml
- Altezza:108 mm
- Seq:03