BOD bottles, WHEATON®
BOD bottles, borosilicate glass, 43×115 mm (Ø×H), number sequence: 73-108, 60 ml
Codice catalogo WHEA227494-03G
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Made of Type I borosilicate glass.
- Available with or without stopper
- With white labeling area
Specifiche
- Capacità:60 ml
- Descrizione:BOD bottles, borosilicate glass, 43×115 mm (Ø×H), number sequence: 73-108