Contagocce, WHEATON®
13 - 425, rosso
Codice catalogo 215-4029
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifiche
- Filettatura:13 - 425
- Seq:16
- Colore tappo:rosso