Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Strip per PCR da 0,2 ml

Strip da 8 provette e strip di tappi ultra trasparenti, trasparente, 0,2 ml, 10 confezioni da 12
  732-0884
Strip per PCR da 0,2 ml
  732-0884
Strip per PCR da 0,2 ml
Strip per PCR da 0,2 ml

Strip da 8 provette e strip di tappi ultra trasparenti, trasparente, 0,2 ml, 10 confezioni da 12

 
SFr. 312.00
   
1 * 120 ST
 

PP.

  • Strips of 8 and 12 tubes with or without cap strips
  • Suitable for 0,2 ml thermal cycler blocks
  • 0,25 ml maximum tube capacity when capped
  • Can be cut into individual tubes
  • Flat or domed cap strips available
  • Certified free from DNase, RNase and Human DNA

Suitable for use in 96-well V-bottom thermal cycler blocks.

Colour: Clear, unless otherwise stated. Other colours available on request.

 
  • Confezione:
    10 confezioni da 12
  • Capacità:
    0,2 ml
  • Colore:
    trasparente
  • Descrizione:
    Strip da 8 provette e strip di tappi ultra trasparenti
  • Seq:
    30
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.