Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer)
TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer), Bottiglia di vetro, 500 mg
Codice catalogo EURLEMR266500
Nome del fornitore: EUROCLONE
Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer)
Codice catalogo EURLEMR266500
Nome del fornitore: EUROCLONE
Numero CAS:
Vai a
Product Details & Documents
Specifiche
Specifiche
- Tipo di conf.:Bottiglia di vetro
- Conf:500 mg
- Descrizione:TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer)
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



