Orders LinkContactUsLinkComponent
N,N-Dimetilformammide
N,N-Dimetilformammide
1 KIT
Catalog # EURLEMR177025
Flammable
Health
Irritant
Supplier:   EUROCLONE
N,N-Dimetilformammide
Catalog # EURLEMR177025
Supplier:  EUROCLONE
CAS Number:  68-12-2
N,N-Dimetilformammide
N,N-Dimetilformammide
1 KIT
CAS Number:  68-12-2
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents
Synonyms:  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  • CAS Number:68-12-2
  • Formula:HCON(CH₃)₂
  • MDL Number:MFCD00003284
  • Molecular Weight:73.09 g/mol
  • Merck Index:13,03269
  • Boiling Pt:153 °C (1013 hPa)
  • Melting Pt:–61 °C
  • Density:0,949 g/cm³ (20 °C)
  • Flash Pt:57,5 °C (coppa chiusa)
  • Storage Temperature:Ambiente
  • EINECS:200-679-5
  • UN:2265
  • ADR:3,III
  • REACH:01-2119475605-32
Specifications
Specifications
  • Conf:
    1 KIT
Specifications