Gelatin
Bottiglia di vetro, 1 kg
Catalog # EURLEMR142001
Supplier: EUROCLONE
Jump to:
Product Details & Documents
Specifications
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:1 kg
- Cat. No.:EURLEMR142001
- Condizioni di conservazione:Ambient
- Protein/peptide name:Gelatin
- Protein synonyms:Gelatine
- CAS No.:9000-70-8
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



