Buffer, HEPES solution (1 mol/L)
HEPES buffer solution (1 mol/l), bottiglia in vetro, 100 mL
Codice catalogo EURLECM0180D-1
Nome del fornitore: EUROCLONE
Buffer, HEPES solution (1 mol/L)
Codice catalogo EURLECM0180D-1
Nome del fornitore: EUROCLONE
Numero CAS:
Vai a
Product Details & Documents
Specifiche
Specifiche
- Tipo di conf.:bottiglia in vetro
- Conf:100 mL
- Descrizione:HEPES buffer solution (1 mol/l)
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



