Cianocobalamina
Glass bottle for solids, 1 mg
Catalog # OXBTCBL64
Marketsource
Supplier: Oxford Biotechnology
Jump to:
Product Details & Documents
- CAS Number:68-19-9
- Formula:C₆₃H₈₈CoN₁₄O₁₄P
- MDL Number:MFCD00151092
- Molecular Weight:1355.38 g/mol
- Merck Index:12,10152
- Melting Pt:320 °C
- Storage Temperature:Refrigerata
- EINECS:200-680-0
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:1 mg



