PORTA X COMMUTATORE X GG CAMERA OSCURA 1 * 1 pezzi
N° de catalogue 730-1203
Fournisseur: Syngene (a division of the Synoptics group)
PORTA X COMMUTATORE X GG CAMERA OSCURA 1 * 1 pezzi
N° de catalogue 730-1203
Fournisseur: Syngene (a division of the Synoptics group)
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Fournisseur: VWR Chemicals
Sealing film for manual application, Parafilm® M
Fournisseur: HEATHROW
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Fournisseur: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
Fournisseur: VWR Chemicals
Guanti in nitrile, Kimtech™ Purple Nitrile™
Fournisseur: KIMBERLY CLARK
VWR®, Pipette sierologiche, Standard Line
Fournisseur: VWR Collection
VWR®, Salviette per il viso
Fournisseur: VWR Collection
VWR®, Guanti in Nitrile
Fournisseur: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Fournisseur: VWR Chemicals
Strisce reattive pH, MQuant®, Supelco®
Fournisseur: Merck
VWR® NITRILE LIGHT, Guanti in nitrile
Fournisseur: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
Fournisseur: VWR Collection