Gouges, Aesculap®
Lamina gouge, cranked, Capener, 16 mm, 235 mm
Katalognr. AESCFL440R
Anbieter: Aesculap
Direkt zu
Product Details & Documents
Technische Daten
- Tipo:Capener
- Larghezza:16 mm
- Descrizione:Lamina gouge, cranked
- Lunghezza:235 mm
- Seq:53