Micro scissors for endoscopic work, Aesculap® MINOP®
Replacement outer tube for MINOP instruments FF385R - FF389R
Codice catalogo AESCFF433R
Nome del fornitore: Aesculap
Micro scissors for endoscopic work, Aesculap® MINOP®
Codice catalogo AESCFF433R
Nome del fornitore: Aesculap
Vai a
Product Details & Documents
Rigid micro scissors for advanced intraventricular Neuroendoscopy.
- Very delicate
Ø = 2 mm
: Complete instrument consists of a handle, an outer tube, and a jaw part with inner tube. Replacements can be ordered separately.
Specifiche
- Lunghezza:Replacement outer tube for MINOP instruments FF385R - FF389R