Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Micro suture tying forceps, Aesculap®

Micro suture tying forceps, Aesculap®

Curved, 0,5 mm jaws, 180 mm
Catalog # AESCFD287R
Supplier:   Aesculap
Micro suture tying forceps, Aesculap®
Catalog # AESCFD287R
Supplier:  Aesculap
Micro suture tying forceps, Aesculap®

Curved, 0,5 mm jaws, 180 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

These micro suture tying forceps feature a round handle.

Specifications
  • Tipo:
    Curved, 0,5 mm jaws
  • Lunghezza:
    180 mm
  • Seq:
    06