Dilator set, Aesculap®
Dilator, 21 mm
Codice catalogo AESCEM121R
Nome del fornitore: Aesculap
Vai a
Product Details & Documents
Complete set of uterine dilators.
: Consists of 26 dilators, 1 rack for dilators (1 to 17 mm), 1 rack for dilators (18 to 26 mm) and metal case with hinged aluminium lid (330×220×80 mm).
Specifiche
- Descrizione:Dilator, 21 mm
- Seq:22