Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Micro suture tying forceps, Aesculap®

Curved, 0,3 mm jaws, 150 mm
  AESCFD285R
 :   Aesculap
Micro suture tying forceps, Aesculap®
  AESCFD285R
 :  Aesculap
Micro suture tying forceps, Aesculap®

Curved, 0,3 mm jaws, 150 mm

 
 
   
1 * 1 ST
 

These micro suture tying forceps feature a round handle.

 
  • Tipo:
    Curved, 0,3 mm jaws
  • Lunghezza:
    150 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.