Digital Solutions
Applications
Support
About
Switzerland
Orders Link
ContactUsLinkComponent
Pinzette
Forceps, micro, jeweller pattern, Aesculap®
Forceps, micro, jeweller pattern, Aesculap®
Angled tip, very fine (0,3 mm), 110 mm
AESCBD329R
:
Aesculap
Forceps, micro, jeweller pattern, Aesculap®
AESCBD329R
:
Aesculap
Forceps, micro, jeweller pattern, Aesculap®
Angled tip, very fine (0,3 mm), 110 mm
SFr. 54.40
1 * 1 ST
-
+
Tipo:
Angled tip, very fine (0,3 mm)
Lunghezza:
110 mm
Seq:
01
Cat. No.:
AESCBD329R