Forceps for cover glasses
Pinze per vetrini, dritte, in acciaio inox, 105 mm
232-0060
: BOCHEM
Stainless steel or stainless steel with PTFE coating.
- For easy, secure handling of cover glasses
- With 6 mm wide tips
- Can also be used for other laboratory equipment, such as filter discs
- PTFE coated design with particularly smooth surface to prevent contamination and enable simple cleaning
- Descrizione:Pinze per vetrini, dritte, in acciaio inox
- Lunghezza:105 mm



