Wire baskets, square and rectangular
150×150×150 mm
Codice catalogo BOCH10025
Nome del fornitore: BOCHEM
Vai a
Product Details & Documents
These stainless steel baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
- Spot-welded grid 8×8×1 mm
- Reinforced edges
- Very good chemical and temperature resistance
Specifiche
- L×P×A:150×150×150 mm