Container, with lid
10000 ml, 325 mm, 220 mm
Catalog # BOCH8082
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10
- Double walled
- Stackable
- With handle
Specifications
- Ø apertura:220 mm
- Capacità:10000 ml
- Altezza:325 mm
Stainless steel 18/10