Stand bases
4,0 kg, 200×315×8 mm
Catalog # BOCH5037
Supplier: BOCHEM
Jump to:
Product Details & Documents
Steel, varnished.
- With M10 threaded hole
- One adjustable foot
Certifications: DIN 12892
Specifications
- L×P×A:200×315×8 mm
- Peso:4,0 kg